siRNA Details
| Gene Symbol: Fras1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175473.3 | NP_780682.3 | 36, 37 | 5706 | ||
| GenBank mRNA | AJ489280.1 | - | 36, 37 | 5706 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fras1| Gene Symbol: Fras1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175473.3 | NP_780682.3 | 36, 37 | 5706 | ||
| GenBank mRNA | AJ489280.1 | - | 36, 37 | 5706 | ||