siRNA Details
| Gene Symbol: Fbxl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033312.3 | NP_001028484.2 | 3 | 627 | ||
| GenBank mRNA | AK049302.1 | - | 3 | 580 | ||
| AK144757.1 | - | 3 | 628 | |||
| AK147353.1 | - | 2 | 2395 | |||
| AK154354.1 | - | 3 | 607 | |||
| BC115599.2 | - | 3 | 599 | |||
| BC139739.1 | - | 3 | 599 | |||

Fbxl18
Genomic Map