siRNA Details
| Gene Symbol: Klrb1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153094.2 | NP_694734.1 | 2 | 216 | ||
| GenBank mRNA | AK041601.1 | - | 2 | 217 | ||
| AK154278.1 | - | 2 | 203 | |||
| AK170023.1 | - | 2 | 193 | |||
| AY029597.1 | - | 2 | 181 | |||
| BC107177.1 | - | 2 | 197 | |||
| BC107178.1 | - | 2 | 197 | |||
| BC127141.1 | - | 2 | 197 | |||
| BC127142.1 | - | 2 | 193 | 40050668 | ||
| DQ237931.1 | - | 2 | 169 | |||
| HQ713435.1 | - | 2 | 169 | |||

Klrb1f
Genomic Map