siRNA Details
| Gene Symbol: Vmn2r54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081449.2 | NP_001074918.2 | 5 | 2285 | ||
| Gene Symbol: Vmn2r53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104644.1 | NP_001098114.1 | 5 | 2285 | ||
More Information
Set Membership: |
|

Vmn2r54
Genomic Map