siRNA Details
Gene Symbol: Vmn2r53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104644.1 | NP_001098114.1 | 5 | 2211 |
Gene Symbol: Vmn2r54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081449.2 | NP_001074918.2 | 5 | 2211 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
