siRNA Details
| Gene Symbol: Lypd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182785.3 | NP_877586.1 | 4, 5 | 831 | ||
| GenBank mRNA | AK077071.1 | - | 4, 5 | 832 | ||
| BC049744.1 | - | 3, 4 | 631 | 6772266 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lypd4| Gene Symbol: Lypd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182785.3 | NP_877586.1 | 4, 5 | 831 | ||
| GenBank mRNA | AK077071.1 | - | 4, 5 | 832 | ||
| BC049744.1 | - | 3, 4 | 631 | 6772266 | ||