siRNA Details
Gene Symbol: Kctd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146188.1 | NP_666300.1 | 3, 4 | 518 | ||
GenBank mRNA | AK034731.1 | - | 4, 5 | 590 | ||
BC031749.1 | - | 3, 4 | 518 | 4507203 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kctd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146188.1 | NP_666300.1 | 3, 4 | 518 | ||
GenBank mRNA | AK034731.1 | - | 4, 5 | 590 | ||
BC031749.1 | - | 3, 4 | 518 | 4507203 |