siRNA Details
| Gene Symbol: Slc5a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146198.2 | NP_666310.1 | 17 | 2138 | ||
| GenBank mRNA | AK008219.1 | - | 17 | 2193 | ||
| BC031742.1 | - | 17 | 2139 | 4221537 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc5a11| Gene Symbol: Slc5a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146198.2 | NP_666310.1 | 17 | 2138 | ||
| GenBank mRNA | AK008219.1 | - | 17 | 2193 | ||
| BC031742.1 | - | 17 | 2139 | 4221537 | ||