siRNA Details
| Gene Symbol: Zfp768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146202.1 | NP_666314.1 | 2 | 1307 | ||
| GenBank mRNA | AK080841.1 | - | 2 | 1382 | ||
| AK155155.1 | - | 2 | 1419 | |||
| BC026432.1 | - | 2 | 1307 | 4235387 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp768| Gene Symbol: Zfp768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146202.1 | NP_666314.1 | 2 | 1307 | ||
| GenBank mRNA | AK080841.1 | - | 2 | 1382 | ||
| AK155155.1 | - | 2 | 1419 | |||
| BC026432.1 | - | 2 | 1307 | 4235387 | ||