siRNA Details
| Gene Symbol: Cbr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145595.2 | NP_663570.2 | 3 | 461 | ||
| GenBank mRNA | AK031756.1 | - | 2 | 1903 | ||
| AK043313.1 | - | 3 | 286 | |||
| AK150763.1 | - | 3 | 462 | |||
| BC009118.1 | - | 3 | 411 | 3154595 | ||

Cbr4
Genomic Map