siRNA Details
| Gene Symbol: Zfp868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045553.1 | NP_001039018.1 | 5 | 835 | ||
| NM_172754.2 | NP_766342.2 | 5 | 850 | |||
| GenBank mRNA | AK083009.1 | - | 5 | 817 | ||
| AK139223.1 | - | 5 | 836 | |||
| AK168306.1 | - | 5 | 840 | |||
| BC010277.1 | - | 4 | 454 | 3586128 | ||

Zfp868
Genomic Map