siRNA Details
| Gene Symbol: Cdk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194444.2 | NP_919426.2 | 10, 11 | 786 | ||
| NM_194446.2 | NP_919428.1 | 10, 11 | 824 | |||
| GenBank mRNA | AK133918.1 | - | 10, 11 | 779 | ||
| AK144801.1 | - | 10, 11 | 796 | |||
| AK170594.1 | - | 10, 11 | 820 | |||
| BC017131.1 | - | 10, 11 | 761 | 4506458 | ||
| BC023736.1 | - | 10, 11 | 805 | |||
| DQ288857.1 | - | 10, 11 | 823 | |||

Cdk10
Genomic Map