siRNA Details
| Gene Symbol: Cdk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194444.2 | NP_919426.2 | 2 | 104 | ||
| NM_194446.2 | NP_919428.1 | 2 | 142 | |||
| GenBank mRNA | AK133918.1 | - | 2 | 97 | ||
| AK144801.1 | - | 2 | 114 | |||
| AK170594.1 | - | 2 | 114 | |||
| BC017131.1 | - | 2 | 79 | 4506458 | ||
| BC023736.1 | - | 2 | 123 | |||
| BC052096.1 | - | 2 | 111 | |||
| BY010457.1 | - | 2 | 105 | |||
| CJ102037.1 | - | 2 | 104 | |||
| DQ288857.1 | - | 2 | 141 | |||

Cdk10
Genomic Map