siRNA Details
| Gene Symbol: Gm4894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177701.3 | NP_808369.1 | 2 | 139 | ||
| GenBank mRNA | AK039996.1 | - | 2 | 140 | ||
| BC132633.1 | - | 2 | 87 | 40130910 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4894| Gene Symbol: Gm4894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177701.3 | NP_808369.1 | 2 | 139 | ||
| GenBank mRNA | AK039996.1 | - | 2 | 140 | ||
| BC132633.1 | - | 2 | 87 | 40130910 | ||