siRNA Details
| Gene Symbol: 4833427G06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177702.3 | NP_808370.1 | 1 | 317 | ||
| GenBank mRNA | AK132837.1 | - | 1 | 280 | ||
| BC048587.1 | - | 1 | 317 | 6707436 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833427G06Rik| Gene Symbol: 4833427G06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177702.3 | NP_808370.1 | 1 | 317 | ||
| GenBank mRNA | AK132837.1 | - | 1 | 280 | ||
| BC048587.1 | - | 1 | 317 | 6707436 | ||