siRNA Details
Gene Symbol: Fbxw19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177703.3 | NP_808371.1 | 5, 6 | 625 | ||
GenBank mRNA | AK087808.1 | - | 5, 6 | 626 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fbxw19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177703.3 | NP_808371.1 | 5, 6 | 625 | ||
GenBank mRNA | AK087808.1 | - | 5, 6 | 626 |