siRNA Details
| Gene Symbol: Lrp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172784.3 | NP_766372.1 | 4 | 1226 | ||
| GenBank mRNA | AK031994.1 | - | 4 | 1168 | ||
| AK036676.1 | - | 4 | 1209 | |||
| AK048919.1 | - | 4 | 1242 | |||
| AK050544.1 | - | 4 | 1196 | |||
| BC059874.1 | - | 4 | 1047 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK006600.1 | - | 417 | Chr. - - [-] on Build | ||

Lrp11
Genomic Map