siRNA Details
| Gene Symbol: Lrp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172784.3 | NP_766372.1 | 6 | 1493 | ||
| GenBank mRNA | AK031994.1 | - | 6 | 1435 | ||
| AK036676.1 | - | 6 | 1476 | |||
| AK050544.1 | - | 6 | 1463 | |||
| BC059874.1 | - | 6 | 1310 | |||

Lrp11
Genomic Map