siRNA Details
| Gene Symbol: C2cd4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168624.1 | NP_001162095.1 | 2 | 253 | ||
| NM_198614.3 | NP_941016.2 | 2 | 379 | |||
| GenBank mRNA | AK029946.1 | - | 2 | 254 | ||
| BC089008.1 | - | 2 | 361 | 30537363 | ||
| BY268666.1 | - | 2 | 380 | |||

C2cd4c
Genomic Map