siRNA Details
Gene Symbol: Zdhhc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080943.2 | NP_001074412.1 | 2 | 699 | ||
GenBank mRNA | AK080584.1 | - | 1 | 363 | ||
BC128021.1 | - | 2 | 703 | 40136338 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zdhhc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080943.2 | NP_001074412.1 | 2 | 699 | ||
GenBank mRNA | AK080584.1 | - | 1 | 363 | ||
BC128021.1 | - | 2 | 703 | 40136338 |