siRNA Details
| Gene Symbol: Cfdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011801.1 | NP_035931.1 | 5 | 641 | ||
| GenBank mRNA | AB010828.1 | - | 5 | 641 | ||
| AK133989.1 | - | 5 | 659 | |||
| AK160532.1 | - | 5 | 606 | |||
| BC005589.1 | - | 5 | 622 | 3964401 | ||
| Y08219.1 | - | 5 | 640 | |||

Cfdp1
Genomic Map