siRNA Details
| Gene Symbol: Clec5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038604.1 | NP_001033693.1 | 5 | 513 | ||
| NM_021364.2 | NP_067339.1 | 4 | 438 | |||
| GenBank mRNA | AF139769.1 | - | 4 | 408 | ||
| AK036697.1 | - | 4 | 404 | |||
| AK046600.1 | - | 4 | 425 | |||
| AK137482.1 | - | 4 | 439 | |||
| BC104364.1 | - | 5 | 472 | 40049817 | ||
| BC104365.1 | - | 5 | 472 | 40049818 | ||
| CN670295.1 | - | 5 | 475 | |||

Clec5a
Genomic Map