siRNA Details
| Gene Symbol: Dmtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110327.1 | NP_001103797.1 | 17 | 2310 | ||
| NM_011806.3 | NP_035936.3 | 18 | 2520 | |||
| GenBank mRNA | AK029147.1 | - | 18 | 2462 | ||
| AK046201.1 | - | 17 | 2905 | |||
| AK078015.1 | - | 17 | 2215 | |||
| AK160595.1 | - | 17 | 2283 | |||
| BC010272.1 | - | 7 | 880 | |||
| BC045141.1 | - | 17 | 2234 | 6307203 | ||
| BC060109.1 | - | 18 | 2442 | |||
| U70017.1 | - | 18 | 2475 | |||

Dmtf1
Genomic Map