siRNA Details
| Gene Symbol: Dleu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173419.2 | NP_775595.1 | 2 | 623 | ||
| GenBank mRNA | AK082171.1 | - | 3 | 385 | ||
| BC038059.1 | - | 2 | 616 | 4235022 | ||
| CK618986.1 | - | 2 | 623 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dleu7| Gene Symbol: Dleu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173419.2 | NP_775595.1 | 2 | 623 | ||
| GenBank mRNA | AK082171.1 | - | 3 | 385 | ||
| BC038059.1 | - | 2 | 616 | 4235022 | ||
| CK618986.1 | - | 2 | 623 | |||