siRNA Details
| Gene Symbol: Impdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011830.3 | NP_035960.2 | 5 | 410 | ||
| GenBank mRNA | AK002676.1 | - | 5 | 442 | ||
| AK011777.1 | - | 5 | 443 | |||
| AK143505.1 | - | 5 | 451 | |||
| AK146362.1 | - | 5 | 476 | |||
| AK150713.1 | - | 5 | 446 | |||
| AK151233.1 | - | 5 | 446 | |||
| AK151706.1 | - | 5 | 469 | |||
| AK152691.1 | - | 5 | 443 | |||
| AK159822.1 | - | 5 | 443 | |||
| AK160569.1 | - | 5 | 443 | |||
| AK166667.1 | - | 5 | 445 | |||
| AK169129.1 | - | 5 | 476 | |||
| AK169788.1 | - | 5 | 476 | |||
| AK215538.1 | - | 3 | 223 | |||
| BC010314.1 | - | 5 | 410 | 2646679 | ||
| BC052671.1 | - | 5 | 431 | 30058399 | ||
| M33934.1 | - | 5 | 443 | |||
| M98333.1 | - | 5 | 377 | |||

Impdh2
Genomic Map