siRNA Details
| Gene Symbol: Phf20l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081409.1 | NP_001074878.1 | 12 | 1739 | ||
| GenBank mRNA | AK032638.1 | - | 12 | 1754 | ||
| AK040888.1 | - | 3 | 470 | |||
| AK041825.1 | - | 2 | 834 | |||
| AK080555.1 | - | 2 | 1371 | |||
| AK187831.1 | - | 1 | 36 | |||
| BC052212.1 | - | 11 | 1665 | |||
| BC061492.1 | - | 11 | 1605 | |||
| BC082777.1 | - | 12 | 1734 | |||
| BC145689.1 | - | 11 | 1470 | 40130992 | ||

Phf20l1
Genomic Map