siRNA Details
Gene Symbol: Phf20l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081409.1 | NP_001074878.1 | 18 | 2538 | ||
GenBank mRNA | AK041825.1 | - | 6 | 3692 | ||
AK046670.1 | - | 1 | 58 | |||
AK053606.1 | - | 8 | 897 |
