siRNA Details
Gene Symbol: Ago2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153178.4 | NP_694818.3 | 11 | 1557 | ||
GenBank mRNA | AB081472.1 | - | 11 | 1419 | ||
AK220193.1 | - | 8 | 922 | |||
BC023279.1 | - | 3 | 259 | |||
BC024857.1 | - | 4 | 402 | |||
BC056639.1 | - | 1 | 124 | |||
BC064741.1 | - | 6 | 674 | |||
BC096465.1 | - | 11 | 1462 | |||
BC128379.1 | - | 11 | 1410 | 40111876 | ||
BC129922.1 | - | 11 | 1410 | 40111875 |
