siRNA Details
Gene Symbol: Ago2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153178.4 | NP_694818.3 | 13 | 1806 | ||
GenBank mRNA | AB081472.1 | - | 13 | 1668 | ||
AK220193.1 | - | 10 | 1171 | |||
BC023279.1 | - | 5 | 507 | |||
BC024857.1 | - | 6 | 651 | |||
BC056639.1 | - | 3 | 373 | |||
BC064741.1 | - | 8 | 923 | |||
BC096465.1 | - | 13 | 1711 | |||
BC128379.1 | - | 13 | 1659 | 40111876 | ||
BC129922.1 | - | 13 | 1659 | 40111875 |
