siRNA Details
| Gene Symbol: Abce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015751.2 | NP_056566.2 | 17 | 1944 | ||
| GenBank mRNA | AK049299.1 | - | 17 | 1946 | ||
| AK088265.1 | - | 1 | 20 | |||
| AK134738.1 | - | 17 | 1881 | |||
| AK144711.1 | - | 17 | 1876 | |||
| AK147151.1 | - | 17 | 1882 | |||
| AK167130.1 | - | 17 | 1892 | |||
| AK167370.1 | - | 17 | 1882 | |||
| AK167704.1 | - | 17 | 1843 | |||
| BC005422.1 | - | 17 | 1865 | 3489529 | ||
| BC066794.1 | - | 16 | 1647 | |||
| BC066836.1 | - | 16 | 1647 | |||
| CD564438.1 | - | 1 | 39 | |||
| U90446.1 | - | 17 | 1833 | |||

Abce1
Genomic Map