siRNA Details
| Gene Symbol: Scn11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011887.3 | NP_036017.3 | 22 | 3520 | ||
| GenBank mRNA | AB031389.1 | - | 22 | 3578 | ||
| AF118044.1 | - | 23 | 3520 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn11a| Gene Symbol: Scn11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011887.3 | NP_036017.3 | 22 | 3520 | ||
| GenBank mRNA | AB031389.1 | - | 22 | 3578 | ||
| AF118044.1 | - | 23 | 3520 | |||