siRNA Details
| Gene Symbol: Sigirr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023059.3 | NP_075546.2 | 3 | 675 | ||
| GenBank mRNA | AF113795.1 | - | 2 | 192 | ||
| AF239957.1 | - | 2 | 186 | |||
| AK133518.1 | - | 3 | 300 | |||
| AK145889.1 | - | 3 | 678 | |||
| BC010806.1 | - | 2 | 197 | 4240425 | ||
| BC094069.1 | - | 3 | 281 | 4974962 | ||

Sigirr
Genomic Map