siRNA Details
Gene Symbol: Sra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001164406.1 | NP_001157878.1 | 5 | 535 | ||
NM_025291.3 | NP_079567.2 | 5 | 638 | |||
GenBank mRNA | AF092039.1 | - | LIT1243 SRA |
4 | 546 | |
AK007302.1 | - | 5 | 624 | |||
AK010375.1 | - | 5 | 624 | |||
AK154232.1 | - | 5 | 608 | |||
AK180031.1 | - | 2 | 161 | |||
BC026480.1 | - | 3 | 417 | |||
BC048362.1 | - | 5 | 610 | 2646489 | ||
BQ748094.1 | - | 3 | 306 |
