siRNA Details
| Gene Symbol: Sra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025291.3 | NP_079567.2 | 4, 5 | 486 | ||
| GenBank mRNA | AK007302.1 | - | 4, 5 | 472 | ||
| AK010375.1 | - | 4, 5 | 472 | |||
| AK154232.1 | - | 4, 5 | 456 | |||
| BC026480.1 | - | 2, 3 | 265 | |||
| BC048362.1 | - | 4, 5 | 458 | 2646489 | ||
| BQ748094.1 | - | 2, 3 | 154 | |||

Sra1
Genomic Map