siRNA Details
| Gene Symbol: Unc80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175510.3 | NP_780719.2 | 3, 4 | 297 | ||
| GenBank mRNA | AK043914.1 | - | 3, 4 | 316 | ||
| AK044008.1 | - | 3, 4 | 330 | |||
| AK081235.1 | - | 3, 4 | 332 | |||
| BC116429.1 | - | 3, 4 | 330 | |||
| BC116430.2 | - | 3, 4 | 330 | 40104344 | ||
| FJ210934.1 | - | 3, 4 | 297 | |||

Unc80
Genomic Map