siRNA Details
| Gene Symbol: Unc80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175510.3 | NP_780719.2 | 8 | 1057 | ||
| GenBank mRNA | AK044008.1 | - | 8 | 1090 | ||
| AK081235.1 | - | 8 | 1092 | |||
| BC116429.1 | - | 8 | 1090 | |||
| BC116430.2 | - | 7 | 950 | 40104344 | ||
| FJ210934.1 | - | 8 | 1057 | |||

Unc80
Genomic Map