siRNA Details
Gene Symbol: Gpr158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004761.1 | NP_001004761.1 | 2 | 1638 | ||
GenBank mRNA | AB093287.1 | - | 2 | 743 | ||
AK030530.1 | - | 2 | 1634 | |||
AK083234.1 | - | 2 | 1664 | |||
AK086256.1 | - | 2 | 1639 |
