siRNA Details
| Gene Symbol: Zfp68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044747.2 | NP_001038212.1 | 5 | 1637 | ||
| NM_001163797.1 | NP_001157269.1 | 6 | 1727 | |||
| NM_013844.2 | NP_038872.1 | 6 | 1733 | |||
| GenBank mRNA | AB010335.1 | - | 1 | 52 | ||
| AB010362.1 | - | 1 | 149 | |||
| AB024005.1 | - | 6 | 1717 | |||
| AK013868.1 | - | 6 | 1728 | |||
| AK044831.1 | - | 5 | 1607 | |||
| AK143158.1 | - | 1 | 133 | |||
| AK146092.1 | - | 6 | 1694 | |||
| AK153887.1 | - | 6 | 1669 | |||
| BC018171.1 | - | 6 | 1716 | 3989444 | ||

Zfp68
Genomic Map