siRNA Details
| Gene Symbol: Zfp68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044747.2 | NP_001038212.1 | 5 | 1805 | ||
| NM_001163797.1 | NP_001157269.1 | 6 | 1895 | |||
| NM_013844.2 | NP_038872.1 | 6 | 1901 | |||
| GenBank mRNA | AB024005.1 | - | 6 | 1885 | ||
| AK013868.1 | - | 6 | 1896 | |||
| AK044831.1 | - | 5 | 1775 | |||
| AK143158.1 | - | 1 | 301 | |||
| AK146092.1 | - | 6 | 1862 | |||
| AK153887.1 | - | 6 | 1837 | |||
| BC018171.1 | - | 6 | 1884 | 3989444 | ||

Zfp68
Genomic Map