siRNA Details
| Gene Symbol: Slc22a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039371.2 | NP_001034460.2 | 7, 8 | 1409 | ||
| GenBank mRNA | AK029695.1 | - | 4, 5 | 567 | ||
| BC094925.1 | - | 7, 8 | 1298 | |||
| BC152347.1 | - | 2, 3 | 304 | |||

Slc22a15
Genomic Map