siRNA Details
| Gene Symbol: Podn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285956.1 | NP_001272885.1 | 10 | 1498 | ||
| NM_001285958.1 | NP_001272887.1 | 10 | 1394 | |||
| NM_172874.3 | NP_766462.2 | 10 | 1420 | |||
| GenBank mRNA | AK034987.1 | - | 9 | 1951 | ||
| AK161018.1 | - | 10 | 1284 | |||
| AY313606.1 | - | 10 | 1420 | |||
| BC052487.1 | - | 10 | 1245 | |||
| BC064034.1 | - | 5 | 582 | |||
| BC094340.1 | - | 10 | 1498 | 30609151 | ||

Podn
Genomic Map