siRNA Details
| Gene Symbol: Fbxl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159963.1 | NP_001153435.1 | 2 | 321 | ||
| NM_178729.4 | NP_848844.1 | 2 | 321 | |||
| GenBank mRNA | AK029957.1 | - | 2 | 269 | ||
| AK041497.1 | - | 2 | 293 | |||
| AK054009.1 | - | 2 | 291 | |||
| AK082931.1 | - | 2 | 291 | |||
| AK085100.1 | - | 2 | 294 | |||
| AK088067.1 | - | 2 | 288 | |||
| AK150664.1 | - | 2 | 281 | |||
| AK159744.1 | - | 2 | 310 | |||
| AK171361.1 | - | 2 | 275 | |||
| AK183264.1 | - | 2 | 171 | |||
| BC047214.1 | - | 234 | 6466511 | |||
| BC113798.1 | - | 2 | 239 | 40062506 | ||
| BY245949.1 | - | 2 | 327 | |||

Fbxl5
Genomic Map