siRNA Details
| Gene Symbol: Hcar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175520.4 | NP_780729.2 | 1 | 996 | ||
| GenBank mRNA | AK029064.1 | - | 1 | 981 | ||
| AK046700.1 | - | 1 | 997 | |||
| BC141305.1 | - | 1 | 867 | 9055917 | ||
| EU809459.1 | - | 1 | 602 | |||
| EU809460.1 | - | 1 | 578 | |||

Hcar1
Genomic Map