siRNA Details
| Gene Symbol: Elfn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175522.3 | NP_780731.1 | 2 | 1468 | ||
| GenBank mRNA | AK044511.1 | - | 2 | 1469 | ||
| BC059029.1 | - | 1 | 1200 | 6811341 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elfn1| Gene Symbol: Elfn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175522.3 | NP_780731.1 | 2 | 1468 | ||
| GenBank mRNA | AK044511.1 | - | 2 | 1469 | ||
| BC059029.1 | - | 1 | 1200 | 6811341 | ||