siRNA Details
| Gene Symbol: Ssu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175525.3 | NP_780734.1 | 9 | 722 | ||
| GenBank mRNA | AK054244.1 | - | 2 | 165 | ||
| AK085578.1 | - | 9 | 723 | |||
| BC116438.2 | - | 9 | 718 | 40104565 | ||
| BC116439.2 | - | 9 | 718 | 40104566 | ||

Ssu2
Genomic Map