siRNA Details
| Gene Symbol: Zfp791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037745.1 | NP_001032834.1 | 5 | 1517 | ||
| GenBank mRNA | BC063331.1 | - | 1303 | 30295602 | ||
| BC100354.1 | - | 5 | 1517 | 30935270 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp791| Gene Symbol: Zfp791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037745.1 | NP_001032834.1 | 5 | 1517 | ||
| GenBank mRNA | BC063331.1 | - | 1303 | 30295602 | ||
| BC100354.1 | - | 5 | 1517 | 30935270 | ||