siRNA Details
| Gene Symbol: Dpy19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172920.4 | NP_766508.3 | 19 | 1734 | ||
| GenBank mRNA | AK031291.1 | - | 1 | 814 | ||
| AK143845.1 | - | 20 | 1739 | |||
| BC031382.1 | - | 12 | 912 | |||
| BC116782.1 | - | 1 | 302 | |||
| BC116784.1 | - | 1 | 302 | |||

Dpy19l1
Genomic Map