siRNA Details
Gene Symbol: Dpy19l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172920.4 | NP_766508.3 | 20 | 1909 | ||
GenBank mRNA | AK031291.1 | - | 2 | 989 | ||
AK143845.1 | - | 21 | 1914 | |||
BC031382.1 | - | 13 | 1087 | |||
BC116782.1 | - | 2 | 477 | |||
BC116784.1 | - | 2 | 477 |
