siRNA Details
| Gene Symbol: Vmn2r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104615.1 | NP_001098085.1 | 5 | 1638 | ||
| Gene Symbol: Vmn2r6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104619.1 | NP_001098089.1 | 5 | 1638 | ||
| Gene Symbol: Vmn2r7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175674.3 | NP_783605.2 | 5 | 1638 | ||
More Information
Set Membership: |
|

Vmn2r4
Genomic Map