siRNA Details
| Gene Symbol: Fktn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139309.4 | NP_647470.1 | 5, 6 | 896 | ||
| GenBank mRNA | AB077383.1 | - | 6, 7 | 1080 | ||
| AJ511807.1 | - | 6, 7 | 916 | |||
| AK085931.1 | - | 5, 6 | 897 | |||
| AK142744.1 | - | 1056 | ||||
| BC017538.1 | - | 6, 7 | 912 | 4506804 | ||

Fktn
Genomic Map